tert-Butyl ((3R,5R)-5-fluoropiperidin-3-yl)carbamate 98%

Supplier: Ambeed
Warning

A501752-100MG A501752-250MG A501752-1G A501752-5G
77169-334EA 29 USD
77169-334 77169-336 77594-570 77613-584
tert-Butyl ((3R,5R)-5-fluoropiperidin-3-yl)carbamate 98%
tert-Butyl ((3R,5R)-5-fluoropiperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₉FN₂O₂
MDL Number: MFCD23105836
CAS Number: 1363378-07-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR