4-Iodo-2-methoxybenzoic acid 95%
Supplier: Ambeed
|
Warning
|
A500833-100G
A500833-10G
A500833-1G
A500833-25G
A500833-5G
77169-320EA
600.49
USD
77169-320
77169-322
77169-324
77169-326
77169-328
4-Iodo-2-methoxybenzoic acid 95%
4-Iodo-2-methoxybenzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇IO₃ MW: 278.0439 g/mol |
MDL Number:
MFCD07780726 CAS Number: 89942-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...