Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride 98%

Supplier: Ambeed
Warning

A500821-100MG A500821-1G A500821-250MG
77169-314EA 58 USD
77169-314 77169-316 77169-318
Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride 98%
Methyl 2-(pyrrolidin-3-yl)acetate hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₄ClNO₂
MDL Number: MFCD13176516
CAS Number: 1126794-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR