[2,2'-Bipyridine]-4-carbaldehyde 95%

Supplier: Ambeed
Warning

A500338-100MG A500338-250MG
77169-310EA 107.12 USD
77169-310 77169-312
[2,2'-Bipyridine]-4-carbaldehyde 95%
[2,2'-Bipyridine]-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O
MW: 184.19 g/mol
MDL Number: MFCD11042459
CAS Number: 146581-82-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR