Dimethyl bicyclo[1.1.1]pentane-1,3-dicarboxylate 98%

Supplier: Ambeed
Warning

A500140-100MG A500140-1G A500140-250MG A500140-5G
77169-302EA 16 USD
77169-302 77169-304 77169-306 77169-308
Dimethyl bicyclo[1.1.1]pentane-1,3-dicarboxylate 98%
Dimethyl bicyclo[1.1.1]pentane-1,3-dicarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂O₄
MDL Number: MFCD23106317
CAS Number: 115913-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR