Methyl 2-amino-4-methoxybenzo[d]thiazole-6-carboxylate 98%

Supplier: Ambeed
Warning

A499712-100MG A499712-250MG A499712-5G A499712-1G
77197-014EA 12.36 USD
77197-014 77197-016 77197-018 77617-138
Methyl 2-amino-4-methoxybenzo[d]thiazole-6-carboxylate 98%
Methyl 2-amino-4-methoxybenzo[d]thiazole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃S
MW: 238.26 g/mol
MDL Number: MFCD23104009
CAS Number: 955886-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR