1,4-Bis(2-bromoethoxy)benzene 98%

Supplier: Ambeed
Danger

A499407-5G A499407-25G
77197-008EA 25 USD
77197-008 77747-020
1,4-Bis(2-bromoethoxy)benzene 98%
1,4-Bis(2-bromoethoxy)benzene

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₂O₂
MW: 324.00 g/mol
MDL Number: MFCD14705063
CAS Number: 5471-84-1
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR