4-Chloro-7-methyl-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Ambeed
Danger

A499205-100MG A499205-250MG
77197-006EA 40.17 USD
77197-006 77747-019
4-Chloro-7-methyl-1H-pyrrolo[3,2-c]pyridine 97%
4-Chloro-7-methyl-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.60 g/mol
MDL Number: MFCD11845525
CAS Number: 1082040-95-6
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR