Methyl-2-chloro-5-(trifluoromethyl)nicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-5-(trifluoromethyl)nicotinate

A498555-10G A498555-1G A498555-250MG A498555-25G A498555-5G
77196-990EA 75 USD
77196-990 77196-992 77196-994 77196-996 77196-998
Methyl-2-chloro-5-(trifluoromethyl)nicotinate 98%
Methyl-2-chloro-5-(trifluoromethyl)nicotinate

Precaución: for Research Use Only

Formula: C₈H₅ClF₃NO₂
MDL Number: MFCD22559950
CAS Number: 1360934-51-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR