4-Bromo-1-ethoxy-2-ethylbenzene 97%

Supplier: Ambeed
Warning

A498391-100MG A498391-1G A498391-250MG
77196-984EA 44.29 USD
77196-984 77196-986 77196-988
4-Bromo-1-ethoxy-2-ethylbenzene 97%
4-Bromo-1-ethoxy-2-ethylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrO
MW: 229.11 g/mol
MDL Number: MFCD18825259
CAS Number: 749932-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR