Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate 98%

Supplier: Ambeed
Warning

A498277-1G A498277-25G A498277-5G
77169-296EA 28 USD
77169-296 77169-298 77169-300
Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate 98%
Ethyl 3-Oxo-3-(pyrazin-2-yl)propionate

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₃
MDL Number: MFCD04114374
CAS Number: 62124-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR