2-(Piperazin-1-yl)benzo[d]thiazole 97%

Supplier: Ambeed
Warning

A498234-10G A498234-1G A498234-250MG A498234-5G
77169-288EA 512.94 USD
77169-288 77169-290 77169-292 77169-294
2-(Piperazin-1-yl)benzo[d]thiazole 97%
2-(Piperazin-1-yl)benzo[d]thiazole

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃S
MW: 219.30 g/mol
MDL Number: MFCD01417996
CAS Number: 55745-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR