(S)-1-(3,5-Difluorophenyl)ethanol 98%

Supplier: Ambeed
Warning

A497496-100MG A497496-1G A497496-250MG A497496-5G
77196-970EA 8 USD
77196-970 77196-972 77196-974 77196-976
(S)-1-(3,5-Difluorophenyl)ethanol 98%
(S)-1-(3,5-Difluorophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈F₂O
MDL Number: MFCD18072180
CAS Number: 692732-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR