1-Benzyl-4-methyl-1,2,3,6-tetrahydropyridine 98%

Supplier: Ambeed
Warning

A496638-10G A496638-1G A496638-250MG A496638-5G
77196-958EA 565.47 USD
77196-958 77196-960 77196-962 77196-964
1-Benzyl-4-methyl-1,2,3,6-tetrahydropyridine 98%
1-Benzyl-4-methyl-1,2,3,6-tetrahydropyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₇N
MW: 187.281 g/mol
MDL Number: MFCD09030042
CAS Number: 32018-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR