1,1-Dibromo-3,3,3-trifluoroacetone 95%

Supplier: Ambeed
Danger

Synonyms: 3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one

A496400-10G A496400-25G
77196-954EA 11 USD
77196-954 77196-956
1,1-Dibromo-3,3,3-trifluoroacetone 95%
1,1-Dibromo-3,3,3-trifluoroacetone

Precaución: for Research Use Only

Formula: Br₂HCC(O)CF₃
MW: 269.84 g/mol
MDL Number: MFCD00041362
CAS Number: 431-67-4
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR