1-Benzyl-3-methyl-1H-imidazol-3-ium 4-methylbenzenesulfonate 99%
Supplier: Ambeed
|
Warning
|
A495599-25G
A495599-5G
A495599-1G
77196-950EA
80
USD
77196-950
77196-952
77747-015
1-Benzyl-3-methyl-1H-imidazol-3-ium 4-methylbenzenesulfonate 99%
1-Benzyl-3-methyl-1H-imidazol-3-ium 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₈H₂₀N₂O₃S MW: 344.42 g/mol |
MDL Number:
MFCD30179601 CAS Number: 52461-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...