1-(2-Aminoethyl)piperidine 98%

Supplier: Ambeed
Danger

Synonyms: N-(2-Aminoethyl)piperidine

A495446-1G A495446-5G A495446-25G
77196-948EA 6 USD
77196-948 77551-504 77572-572
1-(2-Aminoethyl)piperidine 98%
1-(2-Aminoethyl)piperidine

Precaución: for Research Use Only

Formula: C₇H₁₆N₂
MW: 128.22 g/mol
MDL Number: MFCD00006516
CAS Number: 27578-60-5
UN: 2734
ADR: 8,3 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR