3-Cyclopentylpropionic acid 98%

Supplier: Ambeed
Danger

Synonyms: 3-Cyclopentylpropanoic acid, 3-Cylcopentylpropanoic acid

A494486-100G A494486-10G A494486-25G A494486-500G A494486-5G
77196-928EA 59 USD
77196-928 77196-930 77196-932 77196-934 77196-936
3-Cyclopentylpropionic acid 98%
3-Cyclopentylpropionic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₂
MDL Number: MFCD00001392
CAS Number: 140-77-2
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR