(2-Bromo-5-methylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-methylphenylboronic acid

A494330-100MG A494330-1G A494330-250MG A494330-5G
77196-920EA 16 USD
77196-920 77196-922 77196-924 77196-926
(2-Bromo-5-methylphenyl)boronic acid 97%
(2-Bromo-5-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
MDL Number: MFCD26045274
CAS Number: 1450711-53-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR