2-(2-Bromophenyl)-1H-benzimidazole 96%

Supplier: Ambeed
Warning

A493994-1G A493994-250MG A493994-5G A493994-25G
77169-276EA 22 USD
77169-276 77169-278 77169-280 77892-999
2-(2-Bromophenyl)-1H-benzimidazole 96%
2-(2-Bromophenyl)-1H-benzimidazole

Precaución: for Research Use Only

Formula: C₁₃H₉BrN₂
MDL Number: MFCD00449969
CAS Number: 13275-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR