3-((1H-Pyrrolo[2,3-b]pyridin-4-yl)oxy)-N-(4-((4-ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-4-methylbenzamide 98%

Supplier: Ambeed
Warning

Synonyms: NG25

A493735-10MG A493735-1MG A493735-5MG
77169-270EA 45 USD
77169-270 77169-272 77169-274
3-((1H-Pyrrolo[2,3-b]pyridin-4-yl)oxy)-N-(4-((4-ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-4-methylbenzamide 98%
3-((1H-Pyrrolo[2,3-b]pyridin-4-yl)oxy)-N-(4-((4-ethylpiperazin-1-yl)methyl)-3-(trifluoromethyl)phenyl)-4-methylbenzamide

Precaución: for Research Use Only

Formula: C₂₉H₃₀F₃N₅O₂
MW: 537.58 g/mol
MDL Number: MFCD19443255
CAS Number: 1315355-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR