1,3-Dichloro-2-nitrobenzene 98%

Supplier: Ambeed
Warning

A492879-100G A492879-10G A492879-25G A492879-500G A492879-5G
77196-902EA 47 USD
77196-902 77196-904 77196-906 77196-908 77196-910
1,3-Dichloro-2-nitrobenzene 98%
1,3-Dichloro-2-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO₂
MDL Number: MFCD00456795
CAS Number: 601-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR