AQC 95%

Supplier: Ambeed
Warning

Synonyms: 6-Aminoquinoline N-succinimidyl ester, 2,5-Dioxo-1-pyrrolidinyl 6-Quinolylcarbamate

A492242-100MG A492242-1G A492242-250MG
77169-264EA 126 USD
77169-264 77169-266 77551-500
AQC 95%
AQC

Precaución: for Research Use Only

Formula: C₁₄H₁₁N₃O₄
MDL Number: MFCD01940654
CAS Number: 148757-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR