1-Phenyl-3-(piperidin-2-ylmethyl)imidazolidin-2-one hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A492104-100MG
A492104-1G
A492104-250MG
77196-896EA
95.79
USD
77196-896
77196-898
77196-900
1-Phenyl-3-(piperidin-2-ylmethyl)imidazolidin-2-one hydrochloride 97%
1-Phenyl-3-(piperidin-2-ylmethyl)imidazolidin-2-one hydrochloride
Precaución: for Research Use Only
Formula:
C₁₅H₂₂ClN₃O MW: 295.80 g/mol |
MDL Number:
MFCD18086910 CAS Number: 1257326-24-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...