3-Amino-2-chloroisonicotinonitrile 98%

Supplier: Ambeed
Warning

A491505-1G A491505-250MG A491505-5G
77169-252EA 163.77 USD
77169-252 77169-254 77169-256
3-Amino-2-chloroisonicotinonitrile 98%
3-Amino-2-chloroisonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.56 g/mol
MDL Number: MFCD18802419
CAS Number: 342899-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR