1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Di-tert-butyl 2,2'-(1,4,7,10-tetraazacyclododecane-1,7-diyl)diacetate, DO2A-tert-butyl ester

A491084-1G A491084-5G A491084-250MG
77169-248EA 84 USD
77169-248 77169-250 77684-310
1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane 98%, Ambeed.Inc
1,7-Bis(tert-butoxycarbonylmethyl)-1,4,7,10-tetraazacyclododecane

Precaución: For research use only.

Formula: C₂₀H₄₀N₄O₄
MW: 400.56 g/mol
MDL Number: MFCD09263303
CAS Number: 162148-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR