SRPIN340 ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(2-(Piperidin-1-yl)-5-(trifluoromethyl)phenyl)isonicotinamide

A490626-10MG A490626-1MG A490626-50MG A490626-5MG
77169-238EA 17.51 USD
77169-238 77169-240 77169-242 77169-244
SRPIN340 ≥98%
SRPIN340

Precaución: for Research Use Only

Formula: C₁₈H₁₈F₃N₃O
MW: 349.35 g/mol
MDL Number: MFCD00116244
CAS Number: 218156-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR