(2-Bromo-4-chlorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A490499-250MG A490499-5G A490499-1G
77196-892EA 23 USD
77196-892 77196-894 77594-528
(2-Bromo-4-chlorophenyl)boronic acid 98%
(2-Bromo-4-chlorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrClO₂
MDL Number: MFCD22543675
CAS Number: 1451393-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR