2-Chloro-6-methylamino-benzoic acid methyl ester 97+%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-6-(methylamino)benzoate

A490372-100MG A490372-1G A490372-250MG
77196-886EA 33.99 USD
77196-886 77196-888 77196-890
2-Chloro-6-methylamino-benzoic acid methyl ester 97+%
2-Chloro-6-methylamino-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
MW: 199.634 g/mol
MDL Number: MFCD24387373
CAS Number: 1379595-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR