7H-Pyrrolo[2,3-d]pyrimidine-2,4-diol 98%

Supplier: Ambeed
Warning

A490354-1G A490354-250MG A490354-25G A490354-5G A490354-100G
77169-228EA 21.99 USD
77169-228 77169-230 77169-232 77169-234 77892-987
7H-Pyrrolo[2,3-d]pyrimidine-2,4-diol 98%
7H-Pyrrolo[2,3-d]pyrimidine-2,4-diol

Precaución: for Research Use Only

Formula: C₆H₅N₃O₂
MDL Number: MFCD03092515
CAS Number: 39929-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR