2-Benzyloxy-5-bromopyrimidine 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A489793-5G A489793-1G
77169-218EA 40 USD
77169-218 77684-301
2-Benzyloxy-5-bromopyrimidine 98%, Ambeed.Inc
2-Benzyloxy-5-bromopyrimidine

Precaución: For research use only.

Formula: C₁₁H₉BrN₂O
MDL Number: MFCD07375140
CAS Number: 742058-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR