1,4-Benzenedithiol 95%

Supplier: Ambeed
Danger

Synonyms: Benzene-1,4-dithiol

A489711-100MG A489711-1G A489711-250MG A489711-5G
77196-872EA 7 USD
77196-872 77196-874 77196-876 77196-878
1,4-Benzenedithiol 95%
1,4-Benzenedithiol

Precaución: for Research Use Only

Formula: C₆H₆S₂
MW: 142.25 g/mol
Boiling Pt: 240
MDL Number: MFCD00142541
CAS Number: 624-39-5
UN: 3335
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR