2-Iodophenyl acetate 98%

Supplier: Ambeed
Warning

A489707-1G A489707-25G A489707-5G
77196-866EA 28.84 USD
77196-866 77196-868 77196-870
2-Iodophenyl acetate 98%
2-Iodophenyl acetate

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.0445 g/mol
MDL Number: MFCD07780625
CAS Number: 32865-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR