6-Chloro-3H-indole 97%

Supplier: Ambeed
Warning

A489049-100MG A489049-1G A489049-250MG
77196-858EA 70.04 USD
77196-858 77196-860 77196-862
6-Chloro-3H-indole 97%
6-Chloro-3H-indole

Precaución: for Research Use Only

Formula: C₈H₆ClN
MW: 151.59 g/mol
MDL Number: MFCD13177615
CAS Number: 754156-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR