6-Chloro-2-(trifluoromethyl)quinazolin-4(3H)-one 95%
Supplier: Ambeed
|
Warning
|
A488469-100MG
A488469-1G
A488469-250MG
A488469-5G
77169-206EA
22
USD
77169-206
77169-208
77169-210
77169-212
6-Chloro-2-(trifluoromethyl)quinazolin-4(3H)-one 95%
6-Chloro-2-(trifluoromethyl)quinazolin-4(3H)-one
Precaución: for Research Use Only
Formula:
C₉H₄ClF₃N₂O |
MDL Number:
MFCD19703865 CAS Number: 35982-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...