Methyl-2-oxovalerate 90%

Supplier: Ambeed
Danger

Synonyms: 2-Oxovaleric Acid Methyl Ester, 2-Oxopentanoic Acid Methyl Ester, Methyl 2-oxopentanoate, Methyl 2-Oxopentanoate, Methyl 2-Oxovalerate

A487969-1G A487969-250MG A487969-5G
77169-202EA 55.62 USD
77169-202 77169-204 77613-532
Methyl-2-oxovalerate 90%
Methyl-2-oxovalerate

Precaución: for Research Use Only

Formula: C₆H₁₀O₃
MW: 130.14 g/mol
MDL Number: MFCD10566912
CAS Number: 6376-59-6
UN: 1992
ADR: 3,6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR