1,4-Bis(trimethylsilyl)-1,3-butadiyne 95%

Supplier: Ambeed
Danger

Synonyms: 1,4-Bis(trimethylsilyl)buta-1,3-diyne, 1,4-Bis(trimethylsilyl)butadiyne

A487037-1G A487037-250MG A487037-25G A487037-5G
77196-842EA 23.69 USD
77196-842 77196-844 77196-846 77196-848
1,4-Bis(trimethylsilyl)-1,3-butadiyne 95%
1,4-Bis(trimethylsilyl)-1,3-butadiyne

Precaución: for Research Use Only

Formula: C₁₀H₁₈Si₂
MW: 194.42 g/mol
MDL Number: MFCD00009839
CAS Number: 4526-07-2
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR