Ethyl 2-(3,5-dichlorophenyl)acetate 95%

Supplier: Ambeed
Warning

A486070-100MG A486070-250MG A486070-5G
77169-192EA 37.08 USD
77169-192 77169-194 77169-196
Ethyl 2-(3,5-dichlorophenyl)acetate 95%
Ethyl 2-(3,5-dichlorophenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀Cl₂O₂
MW: 233.09 g/mol
MDL Number: MFCD23098648
CAS Number: 200214-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR