1,2-Diamino-2-methylpropane 97%

Supplier: Ambeed
Danger

Synonyms: 2-Methyl-1,2-propanediamine, 2-Methylpropane-1,2-diamine

A485211-1G A485211-250MG A485211-25G A485211-5G
77196-830EA 21.63 USD
77196-830 77196-832 77196-834 77568-952
1,2-Diamino-2-methylpropane 97%
1,2-Diamino-2-methylpropane

Precaución: For research use only.

Formula: C₄H₁₂N₂
MW: 88.15 g/mol
MDL Number: MFCD00008054
CAS Number: 811-93-8
UN: 2734
ADR: 8,3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR