4-Fluoro-1-nitro-2-(trifluoromethoxy)benzene 95%

Supplier: Ambeed
Danger

A484922-100MG A484922-1G A484922-250MG A484922-5G
77169-184EA 39.14 USD
77169-184 77169-186 77169-188 77684-291
4-Fluoro-1-nitro-2-(trifluoromethoxy)benzene 95%
4-Fluoro-1-nitro-2-(trifluoromethoxy)benzene

Precaución: For research use only.

Formula: C₇H₃F₄NO₃
MW: 225.09 g/mol
MDL Number: MFCD15527446
CAS Number: 123572-65-6
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR