2-(2,4-Dichlorophenyl)propan-2-amine hydrochloride 95%

Supplier: Ambeed
Warning

A484725-100MG A484725-1G A484725-250MG
77169-178EA 36.05 USD
77169-178 77169-180 77169-182
2-(2,4-Dichlorophenyl)propan-2-amine hydrochloride 95%
2-(2,4-Dichlorophenyl)propan-2-amine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₂Cl₃N
MW: 240.55 g/mol
MDL Number: MFCD12914022
CAS Number: 1216497-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR