1-Acetylpyrrolidine-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A484684-100MG A484684-1G A484684-250MG
77169-172EA 121.54 USD
77169-172 77169-174 77169-176
1-Acetylpyrrolidine-2-carbaldehyde 95%
1-Acetylpyrrolidine-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₇H₁₁NO₂
MW: 141.16 g/mol
MDL Number: MFCD11583079
CAS Number: 115859-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR