2,3-Dimethylpyridine 1-oxide 95%

Supplier: Ambeed
Warning

A484401-1G A484401-5G
77169-168EA 16.48 USD
77169-168 77169-170
2,3-Dimethylpyridine 1-oxide 95%
2,3-Dimethylpyridine 1-oxide

Precaución: for Research Use Only

Formula: C₇H₉NO
MW: 123.15 g/mol
MDL Number: MFCD00460842
CAS Number: 22710-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR