Dabrafenib 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: N-(3-(5-(2-Aminopyrimidin-4-yl)-2-(tert-butyl)thiazol-4-yl)-2-fluorophenyl)-2,6-difluorobenzenesulphonamide

A484347-1MG A484347-100MG A484347-5MG A484347-10MG A484347-250MG A484347-5G A484347-1G
77169-166EA 6 USD
77169-166 77613-518 77613-520 77684-285 77684-286 77684-287 77892-975
Dabrafenib 98%, Ambeed.Inc
Dabrafenib

Precaución: For research use only.

Formula: C₂₃H₂₀F₃N₅O₂S₂
MDL Number: MFCD17215684
CAS Number: 1195765-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR