ω-Bromo-2,4-dichloroacetophenone 97%

Supplier: Ambeed
Danger

A482443-100G A482443-10G A482443-5G A482443-1G
77196-824EA 380 USD
77196-824 77196-826 77196-828 77746-990
ω-Bromo-2,4-dichloroacetophenone 97%
ω-Bromo-2,4-dichloroacetophenone

Precaución: for Research Use Only

Formula: C₈H₅BrCl₂O
MDL Number: MFCD00053005
CAS Number: 2631-72-3
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR