tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate 95%

Supplier: Ambeed
Warning

A481543-100MG A481543-50MG
77169-150EA 80 USD
77169-150 77169-152
tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate 95%
tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₁₅N₃O₃
MDL Number: MFCD09952358
CAS Number: 954097-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR