(9H-Fluoren-9-yl)methanamine hydrochloride 96%

Supplier: Ambeed
Warning

A481471-100MG A481471-1G A481471-250MG
77169-144EA 30 USD
77169-144 77169-146 77169-148
(9H-Fluoren-9-yl)methanamine hydrochloride 96%
(9H-Fluoren-9-yl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₁₄ClN
MW: 231.72 g/mol
MDL Number: MFCD06804511
CAS Number: 34221-61-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR