4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)-3-fluorophenoxy)-N-methylpicolinamide hydrochloride ≥98%

Supplier: Ambeed
Warning

Synonyms: Regorafenib Hydrochloride

A481225-10MG A481225-1G A481225-1MG A481225-25MG A481225-50MG A481225-5MG A481225-100MG
77196-802EA 10 USD
77196-802 77196-804 77196-806 77196-808 77196-810 77196-812 77746-988
4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)-3-fluorophenoxy)-N-methylpicolinamide hydrochloride ≥98%
4-(4-(3-(4-Chloro-3-(trifluoromethyl)phenyl)ureido)-3-fluorophenoxy)-N-methylpicolinamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₁H₁₆Cl₂F₄N₄O₃
MDL Number: MFCD22417053
CAS Number: 835621-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR