Ethyl 3-Cyclopropylpyrazole-4-carboxylate 95%

Supplier: Ambeed
Warning

A481070-100MG A481070-250MG A481070-5G A481070-1G
77169-138EA 12 USD
77169-138 77169-140 77169-142 77892-966
Ethyl 3-Cyclopropylpyrazole-4-carboxylate 95%
Ethyl 3-Cyclopropylpyrazole-4-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD17171378
CAS Number: 1246471-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR