9-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)nonanoic acid 97%

Supplier: Ambeed
Warning

A480905-1G A480905-250MG A480905-5G
77196-796EA 141.11 USD
77196-796 77196-798 77196-800
9-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)nonanoic acid 97%
9-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)nonanoic acid

Precaución: for Research Use Only

Formula: C₂₄H₂₉NO₄
MW: 395.49 g/mol
MDL Number: MFCD00273465
CAS Number: 212688-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR